Assay Details
| Gene Symbol: Dhx8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144831.2 | NP_659080.2 | 15 - 16 | 2481 | 74 | |
| GenBank mRNA | AK078552.1 | - | 15 - 16 | 2420 | 74 | |
| AK142226.1 | - | 15 - 16 | 2415 | 74 | ||
| AK160848.1 | - | 422 | 74 | |||
| AK170616.1 | - | 15 - 16 | 2411 | 74 | ||
| AK172600.1 | - | 15 - 16 | 2429 | 74 | ||
| AK220485.1 | - | 15 - 16 | 2458 | 74 | ||
| BC065064.1 | - | 15 - 16 | 2410 | 74 | ||
| BC132445.1 | - | 15 - 16 | 2465 | 40130722 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dhx8
Genomic Map