Assay Details
| Gene Symbol: Dhx8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144831.2 | NP_659080.2 | 1 - 2 | 233 | 141 | |
| GenBank mRNA | AK078552.1 | - | 1 - 2 | 172 | 141 | |
| AK142226.1 | - | 1 - 2 | 167 | 141 | ||
| AK153829.1 | - | 1 - 2 | 234 | 141 | ||
| AK170616.1 | - | 1 - 2 | 160 | 141 | ||
| AK172600.1 | - | 1 - 2 | 173 | 141 | ||
| AK220485.1 | - | 1 - 2 | 192 | 141 | ||
| BB654249.1 | - | 1 - 2 | 234 | 141 | ||
| BC065064.1 | - | 1 - 2 | 162 | 141 | ||
| BC132445.1 | - | 1 - 2 | 217 | 40130722 | 141 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dhx8
Genomic Map