Assay Details
| Gene Symbol: Dhx8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144831.2 | NP_659080.2 | 21 - 22 | 3424 | 78 | |
| GenBank mRNA | AK078552.1 | - | 21 - 22 | 3364 | 79 | |
| AK142226.1 | - | 21 - 22 | 3358 | 78 | ||
| AK160848.1 | - | 1365 | 78 | |||
| AK170616.1 | - | 21 - 22 | 3357 | 78 | ||
| AK172600.1 | - | 21 - 22 | 3372 | 78 | ||
| AK220485.1 | - | 21 - 22 | 3401 | 78 | ||
| BC022656.1 | - | 4 - 5 | 629 | 78 | ||
| BC065064.1 | - | 21 - 22 | 3353 | 78 | ||
| BC132445.1 | - | 21 - 22 | 3408 | 40130722 | 78 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dhx8
Genomic Map