Assay Details
| Gene Symbol: Arl4d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025404.3 | NP_079680.1 | 1 - 2 | 97 | 55 | |
| GenBank mRNA | AK004126.1 | - | 1 - 2 | 78 | 55 | |
| AK013320.1 | - | 1 - 2 | 75 | 55 | ||
| BC016113.1 | - | 1 - 2 | 63 | 4481779 | 55 | |
| BQ830332.1 | - | 1 - 2 | 97 | 55 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK004126.1, AK013320.1, BC016113.1, NM_025404.3 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28075093 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28075053 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28075091 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28075090 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2057528 | AK013320.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2057528 | BC016113.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2057528 | NM_025404.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2020612 | AK013320.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2020612 | BC016113.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2020612 | NM_025404.3 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 140375 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 157252 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P180472 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2020612 | AK013320.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2020612 | BC016113.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2020612 | NM_025404.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2057528 | AK013320.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2057528 | BC016113.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2057528 | NM_025404.3 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P402378 | AK013320.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P402378 | BC016113.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P402378 | NM_025404.3 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P402378 | AK004126.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28075073 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28075093 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28075080 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28075079 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28075087 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28075106 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28075103 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28075098 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28075071 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28075088 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28075094 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28075108 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28075099 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28075077 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28075097 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28075068 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28075085 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28075100 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28075083 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28075105 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28075090 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28075109 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28075065 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28075075 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28075081 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28075069 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P180472 | - | - |
| Whole Mouse Genome G4122A | A_51_P235893 | AF312686.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P402378 | AK013320.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P402378 | BC016113.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P402378 | NM_025404.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P402378 | AK004126.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P112821 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P180472 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09300350 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09300349 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09300347 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28075091 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28075090 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28075093 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28075053 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28075127 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28075075 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28075140 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28075059 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28075065 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28075108 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28075057 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28075106 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28075088 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28075062 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28075114 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28075132 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28075050 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28075149 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28075103 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28075156 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28075117 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28075080 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28075109 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28075142 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28075078 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28075052 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28075125 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28075151 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28075060 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28075100 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28075093 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28075053 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28075068 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28075131 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28075063 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28075090 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28075121 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28075044 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28075112 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28075134 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28075087 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28075073 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28075081 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28075098 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28075129 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28075112 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28075117 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09449027 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28075080 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28075125 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09033681 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09641683 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28075131 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28075109 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09628452 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28075085 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28075119 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28075050 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09030168 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28075053 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09655046 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28075066 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09300345 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28075078 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28075067 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28075114 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28075081 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28075129 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09698675 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28075083 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09101333 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09628438 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09698676 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28075052 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09309993 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28075134 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28075090 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28075135 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28075059 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28075106 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28075127 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28075137 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09628448 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09234673 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28075086 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28075073 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28075075 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28075088 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09628447 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09598388 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09234670 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28075120 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28075071 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09101312 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28075108 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28075139 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28075065 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09300354 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28075068 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28075140 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09097873 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28075132 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28075105 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10381475 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10381476 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1418250_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1450358_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10381474 | - | - |
| Mouse Genome 430 2.0 Array | 1418250_at | - | - |
| Mouse Genome 430 2.0 Array | 1450358_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1450358_at | - | - |
| Mouse Genome 430 2.0 Array | 1418250_PM_at | - | - |
| Mouse Expression Set 430 | 1418250_at | - | - |
| Mouse Expression Set 430 | 1450358_at | - | - |
| Mouse Expression Set 430 | 1418250_PM_at | - | - |
| Mouse Expression Set 430 | 1450358_PM_at | - | - |
| Murine Genome U74v2 Set | 100883_at | - | - |
| Murine Genome U74v2 Set | 100883_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1418250_at | - | - |
| Mouse Genome 430A 2.0 Array | 1418250_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1450358_at | - | - |
| Mouse Genome 430A 2.0 Array | 1450358_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1450358_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1450358_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 6180672 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006180672 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2622358 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2593708 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2622358 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2593708 | - | - |
| mouse-6 v1.1 (Illumina) | scl40873.2_406-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6180672 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl40873.2_406-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6180672 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2622354 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2622354 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2593709 | - | - |

Arl4d
Genomic Map