Assay Details
| Gene Symbol: Actr5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175419.4 | NP_780628.3 | 3 - 4 | 1011 | 70 | |
| GenBank mRNA | AK046312.1 | - | 3 - 4 | 1037 | 70 | |
| AK142980.1 | - | 3 - 4 | 1013 | 70 | ||
| BC052039.1 | - | 3 - 4 | 992 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Actr5
Genomic Map