Assay Details
| Gene Symbol: Actr5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175419.4 | NP_780628.3 | 7 - 8 | 1664 | 59 | |
| GenBank mRNA | AK046312.1 | - | 9 - 10 | 1962 | 59 | |
| AK142980.1 | - | 7 - 8 | 1666 | 59 | ||
| BC052039.1 | - | 6 - 7 | 2226 | 59 | ||
| BC069972.1 | - | 1 - 2 | 412 | 59 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Actr5
Genomic Map