Assay Details
| Gene Symbol: Pdcd6ip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164677.1 | NP_001158149.1 | 3 - 4 | 487 | 105 | |
| NM_001164678.1 | NP_001158150.1 | 3 - 4 | 487 | 105 | ||
| NM_011052.2 | NP_035182.2 | 3 - 4 | 487 | 105 | ||
| GenBank mRNA | AF119955.1 | - | 3 - 4 | 448 | 105 | |
| AJ005073.1 | - | 3 - 4 | 335 | 105 | ||
| AJ005074.1 | - | 3 - 4 | 335 | 105 | ||
| AK029827.1 | - | 3 - 4 | 454 | 105 | ||
| AK031256.1 | - | 3 - 4 | 473 | 105 | ||
| AK033943.1 | - | 3 - 4 | 475 | 105 | ||
| AK129340.1 | - | 3 - 4 | 457 | 105 | ||
| AK145893.1 | - | 3 - 4 | 474 | 105 | ||
| AK147736.1 | - | 3 - 4 | 460 | 105 | ||
| AK167574.1 | - | 3 - 4 | 445 | 105 | ||
| AK169704.1 | - | 3 - 4 | 488 | 105 | ||
| AK177760.1 | - | 1 - 2 | 105 | |||
| BC026823.1 | - | 3 - 4 | 429 | 4038684 | 105 | |
| BC051123.1 | - | 3 - 4 | 338 | 6529850 | 105 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pdcd6ip
Genomic Map