Assay Details
| Gene Symbol: Fbxl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178624.6 | NP_848739.1 | 2 - 3 | 120 | 106 | |
| GenBank mRNA | AK039010.1 | - | 2 - 3 | 120 | 106 | |
| AK044693.1 | - | 2 - 3 | 117 | 106 | ||
| AK045742.1 | - | 2 - 3 | 107 | 106 | ||
| AK089994.1 | - | 2 - 3 | 96 | 106 | ||
| AK141177.1 | - | 2 - 3 | 110 | 106 | ||
| AK163472.1 | - | 2 - 3 | 116 | 106 | ||
| BC096582.1 | - | 2 - 3 | 106 | 3963599 | 106 | |
| BC145665.1 | - | 2 - 3 | 100 | 40130968 | 106 | |
| BC145998.1 | - | 2 - 3 | 100 | 40131301 | 106 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK039010.1, AK044693.1, AK045742.1, AK089994.1, AK141177.1, AK163472.1, BC096582.1, BC145665.1, BC145998.1, NM_178624.6 transcripts.
Set Membership: |
|

Fbxl2
Genomic Map