Assay Details
| Gene Symbol: Ccdc55 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012309.2 | NP_001012309.1 | 1 - 2 | 58 | 71 | |
| GenBank mRNA | AK041865.1 | - | 1 - 2 | 44 | 71 | |
| AK046155.1 | - | 1 - 2 | 44 | 71 | ||
| AK078762.1 | - | 1 - 2 | 44 | 71 | ||
| AK078945.1 | - | 1 - 2 | 45 | 71 | ||
| AK161058.1 | - | 1 - 2 | 59 | 71 | ||
| AK162143.1 | - | 1 - 2 | 45 | 71 | ||
| AK162990.1 | - | 1 - 2 | 51 | 71 | ||
| AK168573.1 | - | 1 - 2 | 44 | 71 | ||
| BC089560.1 | - | 1 - 2 | 91 | 30300075 | 71 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK041865.1, AK046155.1, AK162143.1, AK168573.1, BC089560.1, NM_001012309.2 transcripts.
Set Membership: |
|

Ccdc55
Genomic Map