Assay Details
| Gene Symbol: Ccdc55 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012309.2 | NP_001012309.1 | 2 - 3 | 147 | 77 | |
| GenBank mRNA | AK041865.1 | - | 2 - 3 | 133 | 77 | |
| AK046155.1 | - | 2 - 3 | 133 | 77 | ||
| AK078762.1 | - | 2 - 3 | 133 | 77 | ||
| AK078945.1 | - | 2 - 3 | 134 | 77 | ||
| AK161058.1 | - | 2 - 3 | 148 | 77 | ||
| AK162143.1 | - | 2 - 3 | 134 | 77 | ||
| AK162990.1 | - | 2 - 3 | 140 | 77 | ||
| AK168573.1 | - | 2 - 3 | 133 | 77 | ||
| BC016601.1 | - | 2 - 3 | 122 | 77 | ||
| BC089560.1 | - | 2 - 3 | 180 | 30300075 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ccdc55
Genomic Map