Assay Details
| Gene Symbol: Cox4i1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001293559.1 | NP_001280488.1 | 2 - 3 | 131 | 116 | |
| NM_009941.3 | NP_034071.2 | 2 - 3 | 206 | 116 | ||
| GenBank mRNA | AK012583.1 | - | 2 - 3 | 143 | 116 | |
| AK019276.1 | - | 2 - 3 | 108 | 116 | ||
| AK150447.1 | - | 2 - 3 | 136 | 116 | ||
| AV469576.1 | - | 2 - 3 | 131 | 116 | ||
| BC132269.1 | - | 2 - 3 | 127 | 40130546 | 116 | |
| BC132275.1 | - | 2 - 3 | 127 | 40130552 | 116 | |
| BY250039.1 | - | 2 - 3 | 208 | 116 | ||
| M37829.1 | - | 2 - 3 | 129 | 116 | ||
| X54691.1 | - | 2 - 3 | 131 | 116 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cox4i1
Genomic Map