Assay Details
| Gene Symbol: Emc8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010926.5 | NP_035056.1 | 3 - 4 | 509 | 54 | |
| GenBank mRNA | AF052621.1 | - | 3 - 4 | 511 | 54 | |
| AK042704.1 | - | 3 - 4 | 377 | 54 | ||
| AK076710.1 | - | 5 - 6 | 1378 | 54 | ||
| AK143131.1 | - | 3 - 4 | 470 | 54 | ||
| AK161438.1 | - | 3 - 4 | 485 | 54 | ||
| AK164130.1 | - | 3 - 4 | 510 | 54 | ||
| BC009103.1 | - | 3 - 4 | 465 | 3482942 | 54 | |
More Information
Updated Mapping Information
Set Membership: |
|

Emc8
Genomic Map