Assay Details
| Gene Symbol: Mthfsd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC023782.1 | - | 2 - 3 | 219 | 88 | |
| BC026652.1 | - | 1 - 2 | 118 | 88 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Mthfsd
Genomic Map