Assay Details
| Gene Symbol: 1190005I06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_197988.1 | NP_932105.2 | 2 - 3 | 157 | 64 | |
| GenBank mRNA | AK004494.1 | - | 2 - 3 | 106 | 64 | |
| BC027666.1 | - | 2 - 3 | 157 | 1245153 | 64 | |
| Gene Symbol: Emc8-1190005i06rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BQ746342.1 | - | 2 - 3 | 305 | 64 | |
| CX224265.1 | - | 2 - 3 | 368 | 64 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK004494.1, BC027666.1 transcripts.
Set Membership: |
|

1190005I06Rik
Genomic Map