Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0900012169.mm.1 |
NM_001001144.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900012169.mm.1 |
NM_001103162.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900012169.mm.1 |
TC0900001459.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900012173.mm.1 |
NM_001001144.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900012173.mm.1 |
NM_001103162.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900012173.mm.1 |
TC0900001459.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900012172.mm.1 |
NM_001001144.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900012172.mm.1 |
NM_001103162.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900012172.mm.1 |
TC0900001459.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900012170.mm.1 |
NM_001001144.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900012170.mm.1 |
NM_001103162.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900012170.mm.1 |
TC0900001459.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900012171.mm.1 |
NM_001001144.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900012171.mm.1 |
NM_001103162.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900012171.mm.1 |
TC0900001459.mm.1 |
MTA TC |