Assay Details
| Gene Symbol: Gas8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018855.2 | NP_061343.2 | 1 - 2 | 114 | 73 | |
| GenBank mRNA | AK140135.1 | - | 1 - 2 | 93 | 73 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_018855.2 transcripts.
Set Membership: |
|

Gas8
Genomic Map