Assay Details
| Gene Symbol: Gas8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018855.2 | NP_061343.2 | 4 - 5 | 607 | 70 | |
| GenBank mRNA | AK140135.1 | - | 4 - 5 | 586 | 70 | |
| AY692443.1 | - | 4 - 5 | 606 | 70 | ||
| BC003779.1 | - | 4 - 5 | 522 | 3592695 | 70 | |
| DQ189098.1 | - | 3 - 4 | 503 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gas8
Genomic Map