Assay Details
| Gene Symbol: Gas8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018855.2 | NP_061343.2 | 5 - 6 | 661 | 92 | |
| GenBank mRNA | AK140135.1 | - | 5 - 6 | 640 | 92 | |
| AY692443.1 | - | 5 - 6 | 660 | 92 | ||
| BC003779.1 | - | 5 - 6 | 576 | 3592695 | 92 | |
| DQ189098.1 | - | 4 - 5 | 557 | 92 | ||
| U19859.1 | - | 5 - 6 | 682 | 92 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gas8
Genomic Map