Assay Details
| Gene Symbol: Gas8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018855.2 | NP_061343.2 | 8 - 9 | 1125 | 65 | |
| GenBank mRNA | AK140135.1 | - | 8 - 9 | 1104 | 65 | |
| AY692443.1 | - | 8 - 9 | 1124 | 65 | ||
| BC003779.1 | - | 8 - 9 | 1040 | 3592695 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gas8
Genomic Map