Assay Details
| Gene Symbol: Rpain | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252413.1 | NP_001239342.1 | 4 - 5 | 738 | 64 | |
| NM_001252415.1 | NP_001239344.1 | 4 - 5 | 738 | 64 | ||
| NM_027186.2 | NP_081462.1 | 4 - 5 | 738 | 64 | ||
| GenBank mRNA | AK010289.1 | - | 4 - 5 | 467 | 64 | |
| BC047996.1 | - | 4 - 5 | 725 | 6509910 | 64 | |
| DQ810267.1 | - | 4 - 5 | 431 | 64 | ||
| DQ810268.1 | - | 4 - 5 | 431 | 64 | ||
| DQ810269.1 | - | 4 - 5 | 431 | 64 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Rpain
Genomic Map