Assay Details
Gene Symbol: Pabpn1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007462.1 | NP_001007463.1 | 2 - 3 | 439 | 58 | |
GenBank mRNA | BC141316.1 | - | 2 - 3 | 438 | 9055928 | 58 |
BC141317.1 | - | 2 - 3 | 438 | 9055929 | 58 | |
BC145634.1 | - | 2 - 3 | 483 | 9054202 | 58 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
