Assay Details
| Gene Symbol: Pabpn1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007462.1 | NP_001007463.1 | 3 - 4 | 513 | 107 | |
| GenBank mRNA | BC141316.1 | - | 3 - 4 | 512 | 9055928 | 107 |
| BC141317.1 | - | 3 - 4 | 512 | 9055929 | 107 | |
| BC145634.1 | - | 3 - 4 | 557 | 9054202 | 107 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pabpn1l
Genomic Map