Assay Details
| Gene Symbol: Pabpn1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007462.1 | NP_001007463.1 | 4 - 5 | 612 | 71 | |
| GenBank mRNA | BC141316.1 | - | 4 - 5 | 611 | 9055928 | 71 |
| BC141317.1 | - | 4 - 5 | 611 | 9055929 | 71 | |
| BC145634.1 | - | 4 - 5 | 656 | 9054202 | 71 | |
More Information
Set Membership: |
|

Pabpn1l
Genomic Map