Assay Details
| Gene Symbol: Arrb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271358.1 | NP_001258287.1 | 4 - 5 | 306 | 83 | |
| NM_001271359.1 | NP_001258288.1 | 4 - 5 | 306 | 83 | ||
| NM_001271360.1 | NP_001258289.1 | 4 - 5 | 306 | 83 | ||
| NM_145429.5 | NP_663404.1 | 4 - 5 | 306 | 83 | ||
| GenBank mRNA | AK089542.1 | - | 4 - 5 | 241 | 83 | |
| AK150389.1 | - | 4 - 5 | 283 | 83 | ||
| AK154874.1 | - | 4 - 5 | 226 | 83 | ||
| AK159317.1 | - | 4 - 5 | 234 | 83 | ||
| AK162889.1 | - | 4 - 5 | 243 | 83 | ||
| AK170647.1 | - | 4 - 5 | 253 | 83 | ||
| AK170889.1 | - | 4 - 5 | 216 | 83 | ||
| BC016642.1 | - | 4 - 5 | 188 | 2651372 | 83 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Arrb2
Genomic Map