Assay Details
| Gene Symbol: Arrb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271358.1 | NP_001258287.1 | 9 - 10 | 852 | 62 | |
| NM_001271359.1 | NP_001258288.1 | 9 - 10 | 852 | 62 | ||
| NM_001271360.1 | NP_001258289.1 | 9 - 10 | 852 | 62 | ||
| NM_145429.5 | NP_663404.1 | 9 - 10 | 852 | 62 | ||
| GenBank mRNA | AK089542.1 | - | 9 - 10 | 788 | 62 | |
| AK154874.1 | - | 9 - 10 | 772 | 62 | ||
| AK159317.1 | - | 9 - 10 | 780 | 62 | ||
| AK162889.1 | - | 9 - 10 | 789 | 62 | ||
| AK170647.1 | - | 9 - 10 | 799 | 62 | ||
| AK170889.1 | - | 9 - 10 | 762 | 62 | ||
| AK194092.1 | - | 1 - 2 | 45 | 62 | ||
| BC016642.1 | - | 9 - 10 | 734 | 2651372 | 62 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Arrb2
Genomic Map