Assay Details
| Gene Symbol: Zfp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177565.3 | NP_808233.1 | 1 - 2 | 105 | 122 | |
| GenBank mRNA | AK045753.1 | - | 1 - 2 | 106 | 122 | |
| BC096463.1 | - | 1 - 2 | 87 | 6466366 | 122 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK045753.1, BC096463.1, NM_177565.3 transcripts.
Set Membership: |
|

Zfp3
Genomic Map