Assay Details
| Gene Symbol: Nup88 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083331.2 | NP_001076800.1 | 11 - 12 | 1671 | 111 | |
| NM_001276406.1 | NP_001263335.1 | 11 - 12 | 1671 | 111 | ||
| GenBank mRNA | AJ532593.1 | - | 11 - 12 | 1665 | 111 | |
| AK002643.1 | - | 11 - 12 | 1657 | 111 | ||
| AK028563.1 | - | 11 - 12 | 1661 | 111 | ||
| AK166214.1 | - | 11 - 12 | 1661 | 111 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Nup88
Genomic Map