Assay Details
| Gene Symbol: Ppy | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008918.1 | NP_032944.1 | 2 - 3 | 220 | 62 | |
| GenBank mRNA | AK131852.1 | - | 2 - 3 | 240 | 62 | |
| AK136736.1 | - | 2 - 3 | 240 | 62 | ||
| BC099881.1 | - | 2 - 3 | 215 | 40040669 | 62 | |
| BC100676.1 | - | 2 - 3 | 215 | 40040664 | 62 | |
| BC101946.1 | - | 2 - 3 | 215 | 40040662 | 62 | |
| M18208.1 | - | 2 - 3 | 220 | 62 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
positive regulation of defense response to virus by host
neuropeptide signaling pathway
cell-cell signaling
digestion
feeding behavior
regulation of appetite
mitophagy in response to mitochondrial depolarization
G-protein coupled receptor binding
hormone activity
neuropeptide hormone activity
positive regulation of defense response to virus by host
neuropeptide signaling pathway
cell-cell signaling
digestion
feeding behavior
regulation of appetite
mitophagy in response to mitochondrial depolarization
G-protein coupled receptor binding
hormone activity
neuropeptide hormone activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P460791 | BC101946.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P460791 | M18208.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P460791 | NM_008918.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P460791 | AK131852.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P460791 | AK136736.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P460791 | BC101946.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P460791 | M18208.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P460791 | NM_008918.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P460791 | AK131852.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P460791 | AK136736.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 127203 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P460791 | BC101946.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P460791 | M18208.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P460791 | NM_008918.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P460791 | AK131852.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P460791 | AK136736.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P460791 | BC101946.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P460791 | M18208.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P460791 | NM_008918.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P460791 | AK131852.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P460791 | AK136736.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28077562 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28077560 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28077524 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28077512 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28077515 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28077528 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28077537 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28077518 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28077556 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28077513 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28077546 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28077514 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28077565 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28077519 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28077551 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28077558 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28077539 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28077527 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28077543 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28077541 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28077544 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28077530 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28077548 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28077545 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28077525 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28077529 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28077542 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28077563 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28077520 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28077532 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28077523 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28077554 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28077559 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28077533 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28077536 | - | - |
| Whole Mouse Genome G4122A | A_52_P460791 | BC101946.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P460791 | M18208.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P460791 | NM_008918.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P460791 | AK131852.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P460791 | AK136736.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P460791 | BC101946.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P460791 | M18208.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P460791 | NM_008918.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P460791 | AK131852.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P460791 | AK136736.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P112123 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09061718 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09061706 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09061717 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09061712 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09061723 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09061705 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28077518 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28077529 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28077525 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28077542 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28077524 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28077532 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28077545 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28077515 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28077536 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28077559 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28077546 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28077585 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28077465 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28077544 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28077572 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28077569 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28077599 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28077580 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28077512 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28077527 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28077460 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28077539 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28077591 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28077554 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28077576 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28077584 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28077467 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28077476 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28077556 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28077461 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28077533 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28077473 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28077566 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28077455 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28077565 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28077520 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28077470 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28077534 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28077560 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28077549 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28077574 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28077593 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28077568 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28077457 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28077523 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28077479 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28077582 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28077530 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28077514 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28077562 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28077596 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28077558 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28077474 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28077519 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28077519 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09444822 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28077545 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09444795 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09061693 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09061709 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09444815 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09444809 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28077523 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09061710 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09104372 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28077524 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28077525 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28077548 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09061717 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28077539 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28077541 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28077560 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28077509 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28077565 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28077513 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09061712 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09658566 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28077554 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28077549 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28077515 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28077536 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28077533 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28077520 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28077518 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28077558 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09287880 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09444825 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28077514 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28077551 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10391558 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10391559 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10391560 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10391556 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10391557 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1420440_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10391555 | - | - |
| Mouse Genome 430 2.0 Array | 1420440_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1420440_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 19064_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 19064_at | - | - |
| Affymetrix Murine Genome 6500A Array | Msa11000_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1100035444.mm.1 | NM_008918.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100035444.mm.1 | NM_008918.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100035444.mm.1 | TC1100003822.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100035446.mm.1 | NM_008918.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100035446.mm.1 | NM_008918.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100035446.mm.1 | TC1100003822.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100035448.mm.1 | NM_008918.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100035448.mm.1 | NM_008918.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100035448.mm.1 | TC1100003822.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1420440_PM_at | - | - |
| Mouse Expression Set 430 | 1420440_at | - | - |
| Murine Genome U74v2 Set | 162464_f_at | - | - |
| Murine Genome U74v2 Set | 94148_at | - | - |
| Murine Genome U74v2 Set | 94148_PM_at | - | - |
| Murine Genome U74v2 Set | 162464_PM_f_at | - | - |
| Mouse Genome 430A 2.0 Array | 1420440_at | - | - |
| Mouse Genome 430A 2.0 Array | 1420440_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1420440_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1420440_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 798274 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 2340373 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 2340373 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002340373 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2623587 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2623587 | - | - |
| mouse-6 v1.1 (Illumina) | scl39521.4.1_0-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2340373 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl39521.4.1_0-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2340373 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2856567 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2856567 | - | - |

Ppy
Genomic Map