Assay Details
| Gene Symbol: Slc12a4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001253804.1 | NP_001240733.1 | 14 - 15 | 1932 | 63 | |
| NM_009195.3 | NP_033221.1 | 14 - 15 | 1926 | 63 | ||
| GenBank mRNA | AF047339.1 | - | 14 - 15 | 1850 | 63 | |
| AF121118.1 | - | 14 - 15 | 1917 | 63 | ||
| AK151717.1 | - | 14 - 15 | 1887 | 63 | ||
| AK152762.1 | - | 14 - 15 | 1904 | 63 | ||
| AK153245.1 | - | 14 - 15 | 1904 | 63 | ||
| AK159274.1 | - | 14 - 15 | 1914 | 63 | ||
| AK159331.1 | - | 14 - 15 | 1909 | 63 | ||
| AK159483.1 | - | 14 - 15 | 1882 | 63 | ||
| AK159537.1 | - | 14 - 15 | 1909 | 63 | ||
| AK160034.1 | - | 14 - 15 | 1875 | 63 | ||
| AK160072.1 | - | 14 - 15 | 1909 | 63 | ||
| AK164479.1 | - | 14 - 15 | 1891 | 63 | ||
| AK165874.1 | - | 14 - 15 | 1904 | 63 | ||
| AK166304.1 | - | 14 - 15 | 1885 | 63 | ||
| AK167371.1 | - | 14 - 15 | 1923 | 63 | ||
| AK167444.1 | - | 14 - 15 | 1923 | 63 | ||
| BC066872.1 | - | 14 - 15 | 1856 | 30436525 | 63 | |
More Information
Set Membership: |
|

Slc12a4
Genomic Map