Assay Details
| Gene Symbol: Ranbp10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145824.4 | NP_665823.2 | 1 - 2 | 400 | 72 | |
| GenBank mRNA | AK048772.1 | - | 1 - 2 | 256 | 72 | |
| AK173177.1 | - | 1 - 2 | 213 | 72 | ||
| AY337314.1 | - | 1 - 2 | 401 | 72 | ||
| BC024698.1 | - | 1 - 2 | 245 | 3667000 | 72 | |
| EU281316.1 | - | 1 - 2 | 261 | 72 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC024698.1 transcripts.
Set Membership: |
|

Ranbp10
Genomic Map