Assay Details
| Gene Symbol: Dpep2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001301204.1 | NP_001288133.1 | 3 - 4 | 563 | 114 | |
| NM_001301205.1 | NP_001288134.1 | 2 - 3 | 248 | 114 | ||
| NM_176913.4 | NP_795887.2 | 3 - 4 | 678 | 114 | ||
| GenBank mRNA | AF488552.1 | - | 3 - 4 | 678 | 114 | |
| AK089244.1 | - | 3 - 4 | 549 | 114 | ||
| AK155308.1 | - | 3 - 4 | 514 | 114 | ||
| AK155417.1 | - | 3 - 4 | 518 | 114 | ||
| AK170094.1 | - | 3 - 4 | 515 | 114 | ||
| AK170970.1 | - | 3 - 4 | 515 | 114 | ||
| AK171044.1 | - | 3 - 4 | 515 | 114 | ||
| AK171122.1 | - | 3 - 4 | 515 | 114 | ||
| AK171155.1 | - | 2 - 3 | 228 | 114 | ||
| AK171196.1 | - | 3 - 4 | 515 | 114 | ||
| AK171268.1 | - | 3 - 4 | 515 | 114 | ||
| AK171503.1 | - | 3 - 4 | 515 | 114 | ||
| AK171606.1 | - | 3 - 4 | 531 | 114 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dpep2
Genomic Map