Assay Details
| Gene Symbol: Dpep2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001301204.1 | NP_001288133.1 | 8 - 9 | 1173 | 69 | |
| NM_001301205.1 | NP_001288134.1 | 7 - 8 | 858 | 69 | ||
| GenBank mRNA | AK089244.1 | - | 8 - 9 | 1159 | 69 | |
| AK155099.1 | - | 8 - 9 | 1125 | 69 | ||
| AK155308.1 | - | 8 - 9 | 1124 | 69 | ||
| AK155417.1 | - | 8 - 9 | 1128 | 69 | ||
| AK170094.1 | - | 8 - 9 | 1125 | 69 | ||
| AK170970.1 | - | 8 - 9 | 1125 | 69 | ||
| AK171044.1 | - | 8 - 9 | 1125 | 69 | ||
| AK171122.1 | - | 8 - 9 | 1125 | 69 | ||
| AK171155.1 | - | 7 - 8 | 838 | 69 | ||
| AK171196.1 | - | 8 - 9 | 1125 | 69 | ||
| AK171268.1 | - | 8 - 9 | 1125 | 69 | ||
| AK171503.1 | - | 8 - 9 | 1125 | 69 | ||
| AK171606.1 | - | 8 - 9 | 1141 | 69 | ||
More Information
Set Membership: |
|

Dpep2
Genomic Map