Assay Details
| Gene Symbol: Cdh3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037809.5 | NP_001032898.1 | 2 - 3 | 275 | 111 | |
| GenBank mRNA | AK031265.1 | - | 2 - 3 | 215 | 111 | |
| BC098459.1 | - | 2 - 3 | 30079668 | 111 | ||
| X06340.1 | - | 2 - 3 | 213 | 111 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cdh3
Genomic Map