| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0800021943.mm.1 |
NM_027960.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800021943.mm.1 |
NM_027960.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800021943.mm.1 |
TC0800002846.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800021944.mm.1 |
NM_027960.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800021944.mm.1 |
NM_027960.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800021944.mm.1 |
TC0800002846.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800021942.mm.1 |
NM_027960.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800021942.mm.1 |
NM_027960.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800021942.mm.1 |
TC0800002846.mm.1 |
MTA TC |