Assay Details
| Gene Symbol: Itga2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010575.2 | NP_034705.2 | 14 - 15 | 1609 | 79 | |
| GenBank mRNA | AK048700.1 | - | 9 - 10 | 1117 | 79 | |
| AK140749.1 | - | 9 - 10 | 1261 | 79 | ||
| AK142289.1 | - | 14 - 15 | 1471 | 79 | ||
| AK154619.1 | - | 14 - 15 | 1610 | 79 | ||
| AK156872.1 | - | 13 - 14 | 1791 | 79 | ||
| AK157012.1 | - | 13 - 14 | 1793 | 79 | ||
| BC120493.1 | - | 14 - 15 | 1459 | 40129416 | 79 | |
| BC137570.1 | - | 14 - 15 | 1459 | 8860589 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|

Itga2b
Genomic Map