Assay Details
| Gene Symbol: Itga2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010575.2 | NP_034705.2 | 16 - 17 | 1764 | 169 | |
| GenBank mRNA | AF170316.1 | - | 16 - 17 | 1625 | 169 | |
| AK048700.1 | - | 11 - 12 | 1272 | 169 | ||
| AK140749.1 | - | 11 - 12 | 1416 | 169 | ||
| AK142289.1 | - | 16 - 17 | 1626 | 169 | ||
| AK154619.1 | - | 16 - 17 | 1765 | 169 | ||
| AK156872.1 | - | 15 - 16 | 1946 | 169 | ||
| AK157012.1 | - | 15 - 16 | 1948 | 169 | ||
| BC120493.1 | - | 16 - 17 | 1614 | 40129416 | 169 | |
| BC137570.1 | - | 16 - 17 | 1614 | 8860589 | 169 | |
More Information
Updated Mapping Information
Set Membership: |
|

Itga2b
Genomic Map