Assay Details
| Gene Symbol: Itga2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010575.2 | NP_034705.2 | 21 - 22 | 2362 | 120 | |
| GenBank mRNA | AF170316.1 | - | 21 - 22 | 2223 | 120 | |
| AK048700.1 | - | 16 - 17 | 1870 | 120 | ||
| AK140749.1 | - | 16 - 17 | 2014 | 120 | ||
| AK142289.1 | - | 21 - 22 | 2224 | 120 | ||
| AK154619.1 | - | 21 - 22 | 2363 | 120 | ||
| AK156872.1 | - | 20 - 21 | 2544 | 120 | ||
| AK157012.1 | - | 20 - 21 | 2547 | 120 | ||
| AK158443.1 | - | 21 - 22 | 2214 | 120 | ||
| BC120493.1 | - | 21 - 22 | 2212 | 40129416 | 120 | |
| BC137570.1 | - | 21 - 22 | 2212 | 8860589 | 120 | |
More Information
Set Membership: |
|

Itga2b
Genomic Map