Assay Details
| Gene Symbol: Itga2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010575.2 | NP_034705.2 | 24 - 25 | 2622 | 97 | |
| GenBank mRNA | AF045019.1 | - | 1 - 2 | 86 | 97 | |
| AF170316.1 | - | 24 - 25 | 2483 | 97 | ||
| AK048700.1 | - | 19 - 20 | 2130 | 97 | ||
| AK140749.1 | - | 19 - 20 | 2274 | 97 | ||
| AK142289.1 | - | 24 - 25 | 2484 | 97 | ||
| AK154619.1 | - | 24 - 25 | 2623 | 97 | ||
| AK156872.1 | - | 23 - 24 | 2803 | 97 | ||
| AK157012.1 | - | 23 - 24 | 2807 | 97 | ||
| AK158443.1 | - | 24 - 25 | 2474 | 97 | ||
| AK161727.1 | - | 3 - 4 | 498 | 97 | ||
| BC120493.1 | - | 24 - 25 | 2472 | 40129416 | 97 | |
| BC137570.1 | - | 24 - 25 | 2472 | 8860589 | 97 | |
More Information
Updated Mapping Information
Set Membership: |
|

Itga2b
Genomic Map