Assay Details
| Gene Symbol: Itga2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010575.2 | NP_034705.2 | 5 - 6 | 793 | 72 | |
| GenBank mRNA | AF166384.1 | - | 5 - 6 | 671 | 72 | |
| AF170316.1 | - | 5 - 6 | 654 | 72 | ||
| AK048700.1 | - | 1 - 2 | 148 | 72 | ||
| AK142289.1 | - | 5 - 6 | 655 | 72 | ||
| AK154619.1 | - | 5 - 6 | 794 | 72 | ||
| AK156872.1 | - | 5 - 6 | 656 | 72 | ||
| AK157012.1 | - | 5 - 6 | 655 | 72 | ||
| AK158443.1 | - | 5 - 6 | 640 | 72 | ||
| BC120493.1 | - | 5 - 6 | 643 | 40129416 | 72 | |
| BC137570.1 | - | 5 - 6 | 643 | 8860589 | 72 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK048700.1 transcripts.
Set Membership: |
|

Itga2b
Genomic Map