Assay Details
| Gene Symbol: Itga2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010575.2 | NP_034705.2 | 6 - 7 | 844 | 71 | |
| GenBank mRNA | AF166384.1 | - | 6 - 7 | 722 | 71 | |
| AK048700.1 | - | 2 - 3 | 199 | 71 | ||
| AK140749.1 | - | 2 - 3 | 177 | 72 | ||
| AK142289.1 | - | 6 - 7 | 706 | 71 | ||
| AK154619.1 | - | 6 - 7 | 845 | 71 | ||
| AK156872.1 | - | 6 - 7 | 707 | 71 | ||
| AK157012.1 | - | 6 - 7 | 706 | 71 | ||
| AK158443.1 | - | 6 - 7 | 691 | 71 | ||
| BC120493.1 | - | 6 - 7 | 694 | 40129416 | 71 | |
| BC137570.1 | - | 6 - 7 | 694 | 8860589 | 71 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK048700.1 transcripts.
Set Membership: |
|

Itga2b
Genomic Map