Assay Details
| Gene Symbol: Fam171a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199200.2 | NP_954670.2 | 2 - 3 | 491 | 65 | |
| GenBank mRNA | BC025575.1 | - | 2 - 3 | 467 | 65 | |
| CB182176.1 | - | 1 - 2 | 239 | 65 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fam171a2
Genomic Map