Assay Details
| Gene Symbol: Fam171a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199200.2 | NP_954670.2 | 5 - 6 | 930 | 73 | |
| GenBank mRNA | BC025575.1 | - | 5 - 6 | 906 | 73 | |
| BC064455.1 | - | 2 - 3 | 663 | 6810415 | 73 | |
| CB182176.1 | - | 4 - 5 | 678 | 73 | ||
| CB202938.1 | - | 3 - 4 | 396 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fam171a2
Genomic Map