Assay Details
| Gene Symbol: Rbl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001139516.1 | NP_001132988.1 | 6 - 7 | 915 | 154 | |
| NM_011249.2 | NP_035379.2 | 6 - 7 | 915 | 154 | ||
| GenBank mRNA | AK033402.1 | - | 6 - 7 | 909 | 154 | |
| AK147064.1 | - | 6 - 7 | 916 | 154 | ||
| AK156055.1 | - | 6 - 7 | 894 | 154 | ||
| AK178170.1 | - | 1 - 2 | 113 | 154 | ||
| BC060124.1 | - | 6 - 7 | 882 | 6408844 | 154 | |
| U27177.1 | - | 6 - 7 | 899 | 154 | ||
| U27178.1 | - | 6 - 7 | 899 | 154 | ||
| U33320.1 | - | 6 - 7 | 882 | 154 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rbl1
Genomic Map