| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0200042876.mm.1 |
NM_001139520.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200042876.mm.1 |
NM_001139520.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200042876.mm.1 |
NM_018851.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200042876.mm.1 |
NM_018851.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200042876.mm.1 |
TC0200005013.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200042874.mm.1 |
NM_001139520.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200042874.mm.1 |
NM_001139520.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200042874.mm.1 |
NM_018851.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200042874.mm.1 |
NM_018851.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200042874.mm.1 |
TC0200005013.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200042875.mm.1 |
NM_001139520.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200042875.mm.1 |
NM_001139520.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200042875.mm.1 |
NM_018851.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200042875.mm.1 |
NM_018851.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200042875.mm.1 |
TC0200005013.mm.1 |
MTA TC |