Assay Details
| Gene Symbol: Samhd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001139520.1 | NP_001132992.1 | 2 - 3 | 412 | 84 | |
| NM_018851.3 | NP_061339.3 | 2 - 3 | 412 | 84 | ||
| GenBank mRNA | AK054490.1 | - | 2 - 3 | 311 | 84 | |
| AK078936.1 | - | 2 - 3 | 355 | 84 | ||
| AK149865.1 | - | 2 - 3 | 287 | 84 | ||
| AK150964.1 | - | 2 - 3 | 127 | 84 | ||
| AK151335.1 | - | 2 - 3 | 286 | 84 | ||
| AK152815.1 | - | 2 - 3 | 127 | 84 | ||
| AK153390.1 | - | 2 - 3 | 286 | 84 | ||
| AK170688.1 | - | 2 - 3 | 311 | 84 | ||
| AK171565.1 | - | 2 - 3 | 311 | 84 | ||
| BC012721.1 | - | 2 - 3 | 349 | 4037046 | 84 | |
| BC067198.1 | - | 2 - 3 | 335 | 6826682 | 84 | |
| U15635.1 | - | 2 - 3 | 336 | 84 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK150964.1 transcripts.
Set Membership: |
|

Samhd1
Genomic Map