Assay Details
| Gene Symbol: Samhd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001139520.1 | NP_001132992.1 | 7 - 8 | 991 | 57 | |
| NM_018851.3 | NP_061339.3 | 7 - 8 | 991 | 57 | ||
| GenBank mRNA | AK054490.1 | - | 7 - 8 | 890 | 57 | |
| AK078936.1 | - | 7 - 8 | 934 | 57 | ||
| AK128954.1 | - | 6 - 7 | 789 | 57 | ||
| AK149865.1 | - | 7 - 8 | 866 | 57 | ||
| AK151335.1 | - | 7 - 8 | 865 | 57 | ||
| AK153390.1 | - | 7 - 8 | 865 | 57 | ||
| AK170688.1 | - | 7 - 8 | 890 | 57 | ||
| AK171565.1 | - | 7 - 8 | 890 | 57 | ||
| BC012721.1 | - | 7 - 8 | 928 | 4037046 | 57 | |
| BC067198.1 | - | 7 - 8 | 914 | 6826682 | 57 | |
| U15635.1 | - | 7 - 8 | 915 | 57 | ||
More Information
Set Membership: |
|

Samhd1
Genomic Map