Assay Details
| Gene Symbol: Cpm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027468.1 | NP_081744.1 | 3 - 4 | 289 | 76 | |
| GenBank mRNA | BC047389.1 | - | 2 - 3 | 275 | 76 | |
| BC055073.1 | - | 3 - 4 | 294 | 76 | ||
| BC088735.1 | - | 3 - 4 | 294 | 76 | ||
| BC100404.1 | - | 3 - 4 | 283 | 30471377 | 76 | |
More Information
Set Membership: |
|

Cpm
Genomic Map