Assay Details
| Gene Symbol: Cpm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027468.1 | NP_081744.1 | 7 - 8 | 970 | 93 | |
| GenBank mRNA | AK017670.1 | - | 1 - 2 | 200 | 93 | |
| BC047389.1 | - | 6 - 7 | 956 | 93 | ||
| BC055073.1 | - | 7 - 8 | 975 | 93 | ||
| BC088735.1 | - | 7 - 8 | 975 | 93 | ||
| BC100404.1 | - | 7 - 8 | 964 | 30471377 | 93 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cpm
Genomic Map