Assay Details
| Gene Symbol: Dvl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007888.3 | NP_031914.3 | 4 - 5 | 731 | 108 | |
| GenBank mRNA | AK050614.1 | - | 4 - 5 | 535 | 108 | |
| AK144001.1 | - | 4 - 5 | 761 | 108 | ||
| AK146822.1 | - | 4 - 5 | 731 | 108 | ||
| AK159895.1 | - | 4 - 5 | 564 | 108 | ||
| AK168376.1 | - | 4 - 5 | 639 | 108 | ||
| BC021015.1 | - | 4 - 5 | 746 | 108 | ||
| BC053050.1 | - | 4 - 5 | 645 | 6402000 | 108 | |
| BC092396.1 | - | 738 | 30363483 | 108 | ||
| U24160.1 | - | 4 - 5 | 743 | 108 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Dvl2
Genomic Map