Assay Details
| Gene Symbol: Dvl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007888.3 | NP_031914.3 | 8 - 9 | 1163 | 71 | |
| GenBank mRNA | AK050614.1 | - | 8 - 9 | 967 | 71 | |
| AK144001.1 | - | 8 - 9 | 1194 | 71 | ||
| AK146822.1 | - | 8 - 9 | 1163 | 71 | ||
| AK159895.1 | - | 8 - 9 | 996 | 71 | ||
| AK168376.1 | - | 8 - 9 | 1071 | 71 | ||
| BC053050.1 | - | 8 - 9 | 1077 | 6402000 | 71 | |
| U24160.1 | - | 8 - 9 | 1175 | 71 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Dvl2
Genomic Map